Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040433.1 | NP_001035523.1 | 2, 3 | 137 | |
XM_017009709.1 | XP_016865198.1 | 2, 3 | 677 | ||
GenBank mRNA | AY396740.1 | - | 2, 3 | 137 | |
BC137530.1 | - | 2, 3 | 108 | 9021530 | |
BC137531.1 | - | 2, 3 | 108 | 9021531 |