Gene Transcripts

Gene Symbol
MS4A4E
Entrez Gene ID
643680
Gene Name
membrane spanning 4-domains A4E
Gene Aliases
Chromosome Location
Chr. 11: 60201253 - 60243124 on Build GRCh38
UniGene ID
Hs.559934
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011545416.2 XP_011543718.1 3 346
XM_011545417.2 XP_011543719.1 3 346
XM_017018641.1 XP_016874130.1 3 346
GenBank mRNA AB231725.1 - 2 158
AB231729.1 - 2 152
AB231730.1 - 2 158
AB231731.1 - 2 158
AK300102.1 - 3 310

View Full Product Details