Gene Transcripts

Gene Symbol
NAIP
Entrez Gene ID
4671
Gene Name
NLR family apoptosis inhibitory protein
Gene Aliases
BIRC1, NLRB1, psiNAIP
Chromosome Location
Chr. 5: 70968483 - 71025114 on Build GRCh38
UniGene ID
Hs.646951
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004536.2 NP_004527.2 17 4817
NM_022892.1 NP_075043.1 15 4146
XM_011543413.2 XP_011541715.1 7 2955
XM_017009490.1 XP_016864979.1 7 3502
GenBank mRNA AB209123.1 - 6 4203
AK124511.1 - 6 3498
AK295115.1 - 6 2045
BC136273.1 - 15 4154 9020260
BC143761.1 - 14 3986 9052275
U19251.1 - 17 4390

View Full Product Details