Gene Transcripts

Gene Symbol
EYS
Entrez Gene ID
346007
Gene Name
eyes shut homolog (Drosophila)
Gene Aliases
C6orf178, C6orf179, C6orf180, EGFL10, EGFL11, RP25, SPAM, bA166P24.2, bA307F22.3, bA74E24.1, dJ1018A4.2, dJ22I17.2, dJ303F19.1
Chromosome Location
Chr. 6: 63719980 - 65707225 on Build GRCh38
UniGene ID
Hs.25067
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142800.1 NP_001136272.1 15, 16 2922
NM_001292009.1 NP_001278938.1 15, 16 2922
GenBank mRNA FJ416331.1 - 15, 16 2930
FM209056.1 - 15, 16 2874

View Full Product Details