Gene Transcripts

Gene Symbol
COL6A4P1
Entrez Gene ID
344875
Gene Name
collagen type VI alpha 4 pseudogene 1
Gene Aliases
COL6A4, COL6A4P, DIVA, DVWA, VWA6
Chromosome Location
Chr. 3: 15165362 - 15205959 on Build GRCh38
UniGene ID
Hs.722756
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027927.1 2 666
GenBank mRNA AB299979.1 - 2 666
FN394065.1 - 2 500

View Full Product Details