Gene Transcripts

Gene Symbol
Crcp
Entrez Gene ID
12909
Gene Name
calcitonin gene-related peptide-receptor component protein
Gene Aliases
AL022669
Chromosome Location
Chr. 5: 130029306 - 130060787 on Build GRCm38
UniGene ID
Mm.18072
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007761.2 NP_031787.1 4 255
GenBank mRNA AF028242.1 - 4 245
AF118271.1 - 4 255
AK075587.1 - 4 250
BC012676.1 - 4 189 3989860

View Full Product Details