Gene Transcripts

Gene Symbol
Crygb
Entrez Gene ID
12965
Gene Name
crystallin, gamma B
Gene Aliases
Cryg-3, DGcry-3, Nop
Chromosome Location
Chr. 1: 65080222 - 65082290 on Build GRCm38
UniGene ID
Mm.440053
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144761.2 NP_658906.1 3 318
GenBank mRNA AK143110.1 - 3 317
BC056455.1 - 3 295 6824341
EF426303.1 - 3 303
K02585.1 - 2 173

View Full Product Details