Gene Transcripts

Gene Symbol
Crygs
Entrez Gene ID
12970
Gene Name
crystallin, gamma S
Gene Aliases
AI327013, Opj, rncat
Chromosome Location
Chr. 16: 22805203 - 22811410 on Build GRCm38
UniGene ID
Mm.6253
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009967.2 NP_034097.1 2 231
GenBank mRNA AF032995.1 - 2 227
AK143060.1 - 2 228
BC056452.1 - 2 207 6824006

View Full Product Details