Gene Transcripts
- Gene Symbol
- Dr1
- Entrez Gene ID
- 13486
- Gene Name
- down-regulator of transcription 1
- Gene Aliases
- 1700121L09Rik, Dr1l, NC2, NC2beta
- Chromosome Location
- Chr. 5: 108268897 - 108280521 on Build GRCm38
- UniGene ID
- Mm.303534
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026106.4 |
NP_080382.2
|
2, 3 |
1070 |
|
GenBank mRNA |
AK007230.1 |
- |
2, 3 |
1045 |
|
AK018040.1 |
- |
2, 3 |
993 |
|
AK031057.1 |
- |
2, 3 |
1004 |
|
AK075713.1 |
- |
2, 3 |
1070 |
|
AK090113.1 |
- |
2, 3 |
1052 |
|
AK139866.1 |
- |
2, 3 |
1000 |
|
AK144749.1 |
- |
2, 3 |
1064 |
|
AK146573.1 |
- |
2, 3 |
1033 |
|
AK178588.1 |
- |
2, 3 |
290 |
|
AK211344.1 |
- |
2, 3 |
290 |
|
BC013461.1 |
- |
2, 3 |
639 |
4222118 |
View Full Product Details