Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289919.1 | NP_001276848.1 | 5, 6 | 603 | |
NM_010088.2 | NP_034218.1 | 5, 6 | 606 | ||
GenBank mRNA | AF011385.1 | - | 5, 6 | 590 | |
AF015729.1 | - | 5, 6 | 600 | ||
AK131668.1 | - | 5, 6 | 604 | ||
AK146049.1 | - | 5, 6 | 607 | ||
BC061150.1 | - | 5, 6 | 603 | 30317747 |