Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010174.1 | NP_034304.1 | 3 | 306 | |
GenBank mRNA | AK142156.1 | - | 3 | 344 | |
BC002082.1 | - | 3 | 336 | 3485639 | |
BC089542.1 | - | 3 | 344 | 30300963 | |
U02883.1 | - | 3 | 306 |