Gene Transcripts

Gene Symbol
Lect1
Entrez Gene ID
16840
Gene Name
leukocyte cell derived chemotaxin 1
Gene Aliases
Bricd3, ChM-I
Chromosome Location
Chr. 14: 79637682 - 79662199 on Build GRCm38
UniGene ID
Mm.46561
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010701.2 NP_034831.2 6 882
GenBank mRNA AK013975.1 - 6 808
BC045152.1 - 6 781 6439864
U43509.1 - 6 883

View Full Product Details