Gene Transcripts

Gene Symbol
Sh2b3
Entrez Gene ID
16923
Gene Name
SH2B adaptor protein 3
Gene Aliases
AI429800, Lnk
Chromosome Location
Chr. 5: 121815481 - 121837655 on Build GRCm38
UniGene ID
Mm.65493
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK028796.1 - 6 1296
AK156499.1 - 5 1175
AK157297.1 - 5 1165
AK165124.1 - 4 404
AK171128.1 - 6 1239
AK172551.1 - 5 1165
AK172573.1 - 5 1169
BC006759.1 - 5 1149 3709861
U89992.2 - 1060

View Full Product Details