Gene Transcripts

Gene Symbol
Anapc1
Entrez Gene ID
17222
Gene Name
anaphase promoting complex subunit 1
Gene Aliases
2610021O03Rik, AI047775, AI853536, AW547281, Apc1, Mcpr, tsg24
Chromosome Location
Chr. 2: 128610083 - 128687410 on Build GRCm38
UniGene ID
Mm.277408
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008569.2 NP_032595.2 46 5756
GenBank mRNA AK034858.1 - 14 1287
AK090134.1 - 46 5738
BC139002.1 - 46 5737 8862024
BC145492.1 - 46 5737 9054055
X80169.1 - 46 5741

View Full Product Details