Gene Transcripts

Gene Symbol
Mup3
Entrez Gene ID
17842
Gene Name
major urinary protein 3
Gene Aliases
MUP15, MUPIII, Mup-3, Mup25
Chromosome Location
Chr. 4: 62083476 - 62087370 on Build GRCm38
UniGene ID
Mm.250267
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039544.1 NP_001034633.1 3 346
GenBank mRNA AI046324.1 - 3 404
BC019965.1 - 3 308 4194688
BC059102.1 - 3 323 4195272
BC106100.1 - 3 312 5099753
EU882235.1 - 3 265
M27608.1 - 3 295
X03525.1 - 3 294

View Full Product Details