Gene Transcripts

Gene Symbol
Nrl
Entrez Gene ID
18185
Gene Name
neural retina leucine zipper gene
Gene Aliases
D14H14S46E
Chromosome Location
Chr. 14: 55518978 - 55524981 on Build GRCm38
UniGene ID
Mm.20422
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136074.2 NP_001129546.1 3 254
NM_001271916.1 NP_001258845.1 3 274
NM_001271917.1 NP_001258846.1 2 147
NM_008736.3 NP_032762.1 2 253
GenBank mRNA AK044244.1 - 3 275
AK044275.1 - 2 218
AK044316.1 - 3 237
BC031440.1 - 3 238 4527590
BC051241.1 - 1 2085
L14935.1 - 2 192

View Full Product Details