Gene Transcripts

Gene Symbol
Oit3
Entrez Gene ID
18302
Gene Name
oncoprotein induced transcript 3
Gene Aliases
EF-9, LZP
Chromosome Location
Chr. 10: 59422960 - 59441779 on Build GRCm38
UniGene ID
Mm.123013
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010959.2 NP_035089.1 2 249
GenBank mRNA AF356506.1 - 2 196
AF517106.1 - 1 68
AF517107.1 - 1 68
AK040570.1 - 2 250
AK050081.1 - 2 215
AY180915.1 - 2 173

View Full Product Details