Gene Transcripts

Gene Symbol
Olfr23
Entrez Gene ID
18321
Gene Name
olfactory receptor 23
Gene Aliases
MOR135-27, MTPCR50
Chromosome Location
Chr. 11: 73930360 - 73941225 on Build GRCm38
UniGene ID
Mm.246529
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010970.1 NP_035100.1 1 770
GenBank mRNA X89688.1 - 1 396

View Full Product Details