Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011130.2 | NP_035260.1 | 7 | 467 | |
GenBank mRNA | AF513911.1 | - | 7 | 444 | |
AK077127.1 | - | 7 | 452 | ||
AK146745.1 | - | 7 | 456 | ||
AK151436.1 | - | 7 | 452 | ||
AV488131.1 | - | 7 | 465 | ||
BC006681.1 | - | 6 | 415 | ||
BC060998.1 | - | 7 | 448 | 6774075 |