Gene Transcripts
- Gene Symbol
- Ptgdr
- Entrez Gene ID
- 19214
- Gene Name
- prostaglandin D receptor
- Gene Aliases
- DP, PGD
- Chromosome Location
- Chr. 14: 44851235 - 44859444 on Build GRCm38
- UniGene ID
- Mm.5105
- Species
- Mus musculus
- Gene Symbol
- Gm16976
- Entrez Gene ID
- 102637302
- Gene Name
- predicted gene, 16976
- Gene Aliases
- Chromosome Location
- Chr. 14: 44858954 - 44974953 on Build GRCm38
- UniGene ID
- Mm.488934
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK138521.1 |
- |
3 |
729 |
|
View Full Product Details