Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011246.2 | NP_035376.1 | 3 | 472 | |
GenBank mRNA | AF106070.1 | - | 3 | 316 | |
AK013548.1 | - | 3 | 518 | ||
AK134005.1 | - | 3 | 486 | ||
AK141524.1 | - | 3 | 597 | ||
BC057120.1 | - | 3 | 472 | 6831137 | |
BC057341.1 | - | 3 | 472 | 6831137 |