Gene Transcripts

Gene Symbol
Rem1
Entrez Gene ID
19700
Gene Name
rad and gem related GTP binding protein 1
Gene Aliases
E030011C07Rik, Ras, Rem
Chromosome Location
Chr. 2: 152627008 - 152635191 on Build GRCm38
UniGene ID
Mm.8752
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009047.5 NP_033073.1 2 536
GenBank mRNA AK053133.1 - 2 537
AK084460.1 - 2 544
BC150768.1 - 1 388 9087679
U91601.1 - 2 514

View Full Product Details