Gene Transcripts

Gene Symbol
Ring1
Entrez Gene ID
19763
Gene Name
ring finger protein 1
Gene Aliases
Ring1A
Chromosome Location
Chr. 17: 34020792 - 34024680 on Build GRCm38
UniGene ID
Mm.20343
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK155509.1 - 4 540
BC009070.1 - 4 527 2651194
CT010219.1 - 2 254

View Full Product Details