Gene Transcripts

Gene Symbol
Rps16
Entrez Gene ID
20055
Gene Name
ribosomal protein S16
Gene Aliases
AA420385, AI317031
Chromosome Location
Chr. 7: 28350689 - 28352698 on Build GRCm38
UniGene ID
Mm.702
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013647.2 NP_038675.2 5 501
GenBank mRNA AK002904.1 - 1 439
AK010613.1 - 1 442
AK010641.1 - 1 442
AK011060.1 - 1 453
AK019400.2 - 1 440
AK083946.1 - 1 440
AK088413.1 - 1 441
AK088716.1 - 1 441
AK150615.1 - 1 432
AK150808.1 - 1 443
AK168207.1 - 1 440
AK169328.1 - 1 441
BC082286.1 - 1 420
BC090618.1 - 1 468 5713263
BC115673.1 - 1 408 40104131
BC115674.1 - 1 408 40104132

View Full Product Details