Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009274.2 | NP_033300.2 | 9, 10 | 1232 | |
GenBank mRNA | AB006036.1 | - | 9, 10 | 1022 | |
AK136861.1 | - | 9, 10 | 1198 | ||
BC020178.1 | - | 9, 10 | 1129 | 4507346 | |
BC062941.1 | - | 9, 10 | 1151 |