Gene Transcripts

Gene Symbol
Tlx2
Entrez Gene ID
21909
Gene Name
T cell leukemia, homeobox 2
Gene Aliases
Enx, Hox11L.1, Hox11l1, NCX, Ncx1, Tlx1l1, Tlx1l2
Chromosome Location
Chr. 6: 83068325 - 83070225 on Build GRCm38
UniGene ID
Mm.37
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009392.2 NP_033418.1 3 1148
GenBank mRNA AK013072.1 - 2 1224
M75953.1 - 3 1063

View Full Product Details