Gene Transcripts

Gene Symbol
Scmh1
Entrez Gene ID
29871
Gene Name
sex comb on midleg homolog 1
Gene Aliases
AI315320, AI851618, Scml1, Scml3
Chromosome Location
Chr. 4: 120405281 - 120530199 on Build GRCm38
UniGene ID
Mm.427014
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159630.1 NP_001153102.1 10 1311
NM_013883.2 NP_038911.1 10 1311
GenBank mRNA AB030906.1 - 10 1210
AB221680.1 - 6 852
AB221681.1 - 6 852
BC025478.1 - 1 1553
BC034667.1 - 8 1021 5356322

View Full Product Details