Gene Transcripts
- Gene Symbol
- Schip1
- Entrez Gene ID
- 30953
- Gene Name
- schwannomin interacting protein 1
- Gene Aliases
- Nf2ip, Schip-1
- Chromosome Location
- Chr. 3: 68064802 - 68626482 on Build GRCm38
- UniGene ID
- Mm.331064
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001113420.1 |
NP_001106891.1
|
3, 4 |
454 |
|
NM_001113421.1 |
NP_001106892.1
|
4, 5 |
1138 |
|
NM_001282044.1 |
NP_001268973.1
|
4, 5 |
1121 |
|
NM_001282045.1 |
NP_001268974.1
|
4, 5 |
979 |
|
NM_013928.5 |
NP_038956.2
|
3, 4 |
544 |
|
GenBank mRNA |
AF145716.1 |
- |
3, 4 |
650 |
|
AK021361.1 |
- |
3, 4 |
405 |
|
AK083464.1 |
- |
4, 5 |
964 |
|
AK146930.1 |
- |
4, 5 |
1095 |
|
AK168005.1 |
- |
4, 5 |
1048 |
|
BC060529.1 |
- |
3, 4 |
478 |
|
BC094348.1 |
- |
4, 5 |
1047 |
6414097 |
EU163407.1 |
- |
4, 5 |
1138 |
|
EU163408.1 |
- |
3, 4 |
454 |
|
KM233715.1 |
- |
3, 4 |
400 |
|
KM233716.1 |
- |
3, 4 |
405 |
|
- Gene Symbol
- Gm21949
- Entrez Gene ID
- 100505386
- Gene Name
- predicted gene, 21949
- Gene Aliases
- Iqcj-schip1, Schip-1-iqcj, Schip1
- Chromosome Location
- Chr. 3: 67892220 - 68626482 on Build GRCm38
- UniGene ID
- Mm.331064
- Species
- Mus musculus
View Full Product Details