Gene Transcripts

Gene Symbol
Nptx2
Entrez Gene ID
53324
Gene Name
neuronal pentraxin 2
Gene Aliases
Narp, np2
Chromosome Location
Chr. 5: 144545887 - 144557478 on Build GRCm38
UniGene ID
Mm.10099
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016789.3 NP_058069.1 3 843
GenBank mRNA AF049124.1 - 3 733
AK134806.1 - 3 844
BC026054.1 - 3 838 4038070
DV649643.1 - 3 282

View Full Product Details