Gene Transcripts

Gene Symbol
Snrpa
Entrez Gene ID
53607
Gene Name
small nuclear ribonucleoprotein polypeptide A
Gene Aliases
C430021M15Rik, Rnu1a-1, Rnu1a1, U1-A, U1A
Chromosome Location
Chr. 7: 27187006 - 27196271 on Build GRCm38
UniGene ID
Mm.386890
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001046637.1 NP_001040102.1 2, 3 491
NM_001285825.1 NP_001272754.1 3, 4 565
NM_015782.3 NP_056597.3 3, 4 568
GenBank mRNA AK013886.1 - 3, 4 542
AK088550.1 - 3, 4 426
AK206903.1 - 1, 2 26
BC003229.1 - 3, 4 468 3483391
BC094006.1 - 3, 4 428 4983854
BC096648.1 - 3, 4 422 1397390
CX565356.1 - 2, 3 461
DV650448.1 - 1, 2 184
L15447.1 - 3, 4 364

View Full Product Details