Gene Transcripts

Gene Symbol
Rp9
Entrez Gene ID
55934
Gene Name
retinitis pigmentosa 9 (human)
Gene Aliases
PAP-1, Rp9h
Chromosome Location
Chr. 9: 22448311 - 22468356 on Build GRCm38
UniGene ID
Mm.38235
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018739.2 NP_061209.1 3 433
GenBank mRNA AK146136.1 - 3 414
AK147912.1 - 436
AK168391.1 - 3 426
BC048480.1 - 3 291
BC115549.1 - 3 393 40101700
BC115550.1 - 3 393 40101701
BY239981.1 - 3 434
D78255.1 - 3 412

View Full Product Details