Gene Transcripts
- Gene Symbol
- KRT32
- Entrez Gene ID
- 3882
- Gene Name
- keratin 32
- Gene Aliases
- HA2, HKA2, KRTHA2, hHa2
- Chromosome Location
- Chr. 17: 41459513 - 41469252 on Build GRCh38
- UniGene ID
- Hs.41752
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002278.3 |
NP_002269.3
|
2 |
566 |
|
GenBank mRNA |
X81419.1 |
- |
2 |
405 |
|
View Full Product Details