Gene Transcripts

Gene Symbol
Snd1
Entrez Gene ID
56463
Gene Name
staphylococcal nuclease and tudor domain containing 1
Gene Aliases
AL033314, Tudor-SN
Chromosome Location
Chr. 6: 28480315 - 28935162 on Build GRCm38
UniGene ID
Mm.439987
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019776.2 NP_062750.2 10, 11 1367
GenBank mRNA AB021491.1 - 10, 11 1324
AK034914.1 - 9, 10 1236
AK088028.1 - 10, 11 1326
AK133837.1 - 10, 11 1373
AK159837.1 - 10, 11 1332
AK161591.1 - 10, 11 1321
AK162432.1 - 10, 11 1308
AK167576.1 - 10, 11 1357
AK185603.1 - 3, 4 235
AK190012.1 - 3, 4 235
AK196687.1 - 3, 4 235
AK200045.1 - 3, 4 235
AK216953.1 - 3, 4 235
BC007126.1 - 10, 11 1314 3592548

View Full Product Details