Gene Transcripts

Gene Symbol
LAMA5
Entrez Gene ID
3911
Gene Name
laminin subunit alpha 5
Gene Aliases
Chromosome Location
Chr. 20: 62309060 - 62367318 on Build GRCh38
UniGene ID
Hs.473256
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005560.4 NP_005551.3 64 8771
XM_006723796.3 XP_006723859.1 64 8774
XM_011528818.2 XP_011527120.1 63 8636
XM_011528819.2 XP_011527121.1 62 8501
GenBank mRNA AB011105.1 - 19 2556
AF443072.1 - 64 8771
AK074307.1 - 2 85
Z95636.1 - 4 478

View Full Product Details