Gene Transcripts
- Gene Symbol
- Htra2
- Entrez Gene ID
- 64704
- Gene Name
- HtrA serine peptidase 2
- Gene Aliases
- AI481710, Omi, Prss25, mnd2
- Chromosome Location
- Chr. 6: 83051266 - 83054571 on Build GRCm38
- UniGene ID
- Mm.21880
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019752.3 |
NP_062726.3
|
3, 4 |
1044 |
|
GenBank mRNA |
AF164513.1 |
- |
3, 4 |
923 |
|
AF175324.1 |
- |
3, 4 |
940 |
|
AK011536.1 |
- |
|
1850 |
|
AK146338.1 |
- |
3, 4 |
935 |
|
AK159188.1 |
- |
3, 4 |
864 |
|
AK176936.1 |
- |
1, 2 |
27 |
|
AK178298.1 |
- |
1, 2 |
27 |
|
AK182973.1 |
- |
1, 2 |
27 |
|
AK186135.1 |
- |
1, 2 |
27 |
|
AK188221.1 |
- |
1, 2 |
27 |
|
AK215600.1 |
- |
1, 2 |
27 |
|
BC033276.1 |
- |
3, 4 |
904 |
|
BC049880.1 |
- |
3, 4 |
1044 |
5321861 |
View Full Product Details