Gene Transcripts
- Gene Symbol
- Dpy30
- Entrez Gene ID
- 66310
- Gene Name
- dpy-30 homolog (C. elegans)
- Gene Aliases
- 2810410M20Rik, C87842
- Chromosome Location
- Chr. 17: 74299474 - 74323944 on Build GRCm38
- UniGene ID
- Mm.28536
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001146222.1 |
NP_001139694.1
|
3, 4 |
303 |
|
NM_001146223.1 |
NP_001139695.1
|
3, 4 |
217 |
|
NM_001146224.1 |
NP_001139696.1
|
2, 3 |
223 |
|
NM_024428.4 |
NP_077746.3
|
3, 4 |
312 |
|
GenBank mRNA |
AK003688.1 |
- |
2, 3 |
148 |
|
AK011643.1 |
- |
2, 3 |
122 |
|
AK013073.1 |
- |
3, 4 |
206 |
|
AK077902.1 |
- |
3, 4 |
205 |
|
AK214978.1 |
- |
2, 3 |
95 |
|
AV460563.1 |
- |
3, 4 |
218 |
|
AY297538.1 |
- |
3, 4 |
316 |
|
BC002240.1 |
- |
3, 4 |
186 |
3491912 |
BU961422.1 |
- |
3, 4 |
318 |
|
BX518196.1 |
- |
3, 4 |
318 |
|
View Full Product Details