Gene Transcripts

Gene Symbol
Lsm5
Entrez Gene ID
66373
Gene Name
LSM5 homolog, U6 small nuclear RNA associated (S. cerevisiae)
Gene Aliases
2010208O10Rik, 2310034K10Rik
Chromosome Location
Chr. 6: 56701057 - 56704699 on Build GRCm38
UniGene ID
Mm.25642
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025520.3 NP_079796.1 4 256
GenBank mRNA AK008480.1 - 3 514
AK009617.1 - 4 250
AK011218.1 - 4 242
BC048459.1 - 4 231 6601124
BC061085.1 - 240 30295556
BC132626.1 - 4 256 40130903
BC132628.1 - 4 256 40130905
CB204385.1 - 4 249

View Full Product Details