Gene Transcripts

Gene Symbol
MAN2A1
Entrez Gene ID
4124
Gene Name
mannosidase alpha class 2A member 1
Gene Aliases
AMan II, GOLIM7, MANA2, MANII
Chromosome Location
Chr. 5: 109689366 - 109869625 on Build GRCh38
UniGene ID
Hs.432822
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002372.3 NP_002363.2 19 3987
XM_017009472.1 XP_016864961.1 19 2908
GenBank mRNA AK314090.1 - 19 3328
BC142656.1 - 19 3289 40148064
BC142696.1 - 19 3289 40148740
D63998.1 - 19 3445
U31520.1 - 19 2938

View Full Product Details