Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026257.3 | NP_080533.1 | 12 | 1111 | |
GenBank mRNA | AK010492.1 | - | 5 | 590 | |
AK015720.1 | - | 12 | 1095 | ||
AK048494.1 | - | 12 | 1113 | ||
BC019461.1 | - | 11 | 967 | 4224033 |