Gene Transcripts
- Gene Symbol
- Tsr3
- Entrez Gene ID
- 68327
- Gene Name
- TSR3 20S rRNA accumulation
- Gene Aliases
- 0610007P22Rik, 1110014B11Rik, AL022759
- Chromosome Location
- Chr. 17: 25240170 - 25242799 on Build GRCm38
- UniGene ID
- Mm.180687
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163718.1 |
NP_001157190.1
|
3, 4 |
588 |
|
NM_026676.3 |
NP_080952.2
|
3, 4 |
609 |
|
GenBank mRNA |
AK002309.1 |
- |
3, 4 |
581 |
|
AK003675.1 |
- |
1, 2 |
1060 |
|
AK158743.1 |
- |
3, 4 |
610 |
|
AK169072.1 |
- |
2, 3 |
944 |
|
AK170074.1 |
- |
3, 4 |
578 |
|
BC027751.1 |
- |
3, 4 |
679 |
5011894 |
BC038884.1 |
- |
1, 2 |
382 |
1516275 |
BC089019.1 |
- |
3, 4 |
558 |
30551363 |
CA979987.1 |
- |
3, 4 |
537 |
|
View Full Product Details