Gene Transcripts

Gene Symbol
Pmvk
Entrez Gene ID
68603
Gene Name
phosphomevalonate kinase
Gene Aliases
1110011E12Rik, 2900002L22Rik, AV001327, PMK, PMKA, PMKASE
Chromosome Location
Chr. 3: 89454546 - 89469014 on Build GRCm38
UniGene ID
Mm.34242
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026784.3 NP_081060.2 1 297
GenBank mRNA AK003607.1 - 1 100
AK150483.1 - 1 216
AK168358.1 - 1 192
BY266316.1 - 1 299

View Full Product Details