Gene Transcripts

Gene Symbol
Snx5
Entrez Gene ID
69178
Gene Name
sorting nexin 5
Gene Aliases
0910001N05Rik, 1810032P22Rik, AU019504, D2Ertd52e
Chromosome Location
Chr. 2: 144250123 - 144270902 on Build GRCm38
UniGene ID
Mm.273379
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199188.1 NP_001186117.1 12, 13 1323
NM_024225.5 NP_077187.1 11, 12 1234
GenBank mRNA AK007676.1 - 11, 12 1182
AK029463.1 - 11, 12 1194
AK031873.1 - 12, 13 1215
AK049129.1 - 12, 13 1215
AK078776.1 - 12, 13 1216
AK144710.1 - 12, 13 1219
AK152311.1 - 12, 13 1260
AK159115.1 - 8, 9 777
AK159904.1 - 11, 12 1139
AK169545.1 - 10, 11 2129
AK169786.1 - 12, 13 1215
AK210004.1 - 1, 2 102
AK217080.1 - 1, 2 102
BC002242.1 - 12, 13 1292 3492262

View Full Product Details