Gene Transcripts

Gene Symbol
Zpbp2
Entrez Gene ID
69376
Gene Name
zona pellucida binding protein 2
Gene Aliases
1700017D11Rik, 2610022C02Rik, ZPBPL
Chromosome Location
Chr. 11: 98551097 - 98558665 on Build GRCm38
UniGene ID
Mm.55996
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166494.1 NP_001159966.1 3 638
NM_001166495.1 NP_001159967.1 4 427
NM_027061.2 NP_081337.2 4 518
NM_199419.3 NP_955451.2 4 518
GenBank mRNA AK006058.1 - 3 586
AK137467.1 - 2 1592
AY251601.1 - 4 463
AY251602.1 - 4 463
BC050986.1 - 4 394
BC089476.1 - 4 518 6708062

View Full Product Details