Gene Transcripts

Gene Symbol
Plekhm2
Entrez Gene ID
69582
Gene Name
pleckstrin homology domain containing, family M (with RUN domain) member 2
Gene Aliases
2310034J19Rik, AI854247, mKIAA0842
Chromosome Location
Chr. 4: 141625732 - 141664151 on Build GRCm38
UniGene ID
Mm.33220
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033150.1 NP_001028322.1 6 694
GenBank mRNA AK122386.1 - 778
AK156652.1 - 3 316
AK219223.1 - 5 359
BC058984.1 - 4 285

View Full Product Details