Gene Transcripts

Gene Symbol
Ptcd3
Entrez Gene ID
69956
Gene Name
pentatricopeptide repeat domain 3
Gene Aliases
2610034F17Rik, 2810422B04Rik, AA589622, AU045708, MRP-S39
Chromosome Location
Chr. 6: 71880638 - 71908772 on Build GRCm38
UniGene ID
Mm.30256
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027275.3 NP_081551.2 5 303
GenBank mRNA AK032831.1 - 5 287
AK146005.1 - 5 292
AK146172.1 - 5 304
AK172480.1 - 5 291
BC115434.2 - 5 299 40094203

View Full Product Details