Gene Transcripts

Gene Symbol
Pds5a
Entrez Gene ID
71521
Gene Name
PDS5, regulator of cohesion maintenance, homolog A (S. cerevisiae)
Gene Aliases
9030416H16Rik, E230024D05Rik, mKIAA0648
Chromosome Location
Chr. 5: 65608124 - 65698163 on Build GRCm38
UniGene ID
Mm.278974
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081321.1 NP_001074790.1 26 3218
GenBank mRNA AK018509.1 - 2 1144
AK136537.1 - 2 2288
AK172992.1 - 25 4915
BC021408.1 - 13 1601

View Full Product Details