Gene Transcripts

Gene Symbol
Tysnd1
Entrez Gene ID
71767
Gene Name
trypsin domain containing 1
Gene Aliases
1300019N10Rik
Chromosome Location
Chr. 10: 61695497 - 61702773 on Build GRCm38
UniGene ID
Mm.71514
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001272090.1 NP_001259019.1 4 1937
NM_001272091.1 NP_001259020.1 3 1847
NM_001272092.1 NP_001259021.1 3 1792
NM_027912.1 NP_082188.1 4 1978
GenBank mRNA AK005069.1 - 4 1979
AK154513.1 - 4 1961
BC119320.1 - 4 1956 8734069

View Full Product Details