Gene Transcripts

Gene Symbol
Ppm1j
Entrez Gene ID
71887
Gene Name
protein phosphatase 1J
Gene Aliases
2310008J22Rik, PP2Czeta, Ppp2cz
Chromosome Location
Chr. 3: 104781006 - 104786019 on Build GRCm38
UniGene ID
Mm.379204
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027982.2 NP_082258.2 8 1217
GenBank mRNA AK009235.1 - 8 1156
AY184802.1 - 8 1217
BC117498.1 - 8 1213 40106734
BC121788.1 - 8 1212 40106732

View Full Product Details