Gene Transcripts

Gene Symbol
Asb6
Entrez Gene ID
72323
Gene Name
ankyrin repeat and SOCS box-containing 6
Gene Aliases
2510004M11Rik, AA409356
Chromosome Location
Chr. 2: 30823097 - 30828300 on Build GRCm38
UniGene ID
Mm.27656
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133346.2 NP_579924.1 5 658
GenBank mRNA AF398967.1 - 2696
AK010909.1 - 5 639
AK049312.1 - 5 647
AK088890.1 - 5 593
AK190893.1 - 2 136
AY665653.1 - 5 544
BC019192.1 - 5 650 5035395
BC085485.1 - 4 1339

View Full Product Details