Gene Transcripts

Gene Symbol
MYH4
Entrez Gene ID
4622
Gene Name
myosin heavy chain 4
Gene Aliases
MYH2B, MyHC-2B, MyHC-IIb
Chromosome Location
Chr. 17: 10443058 - 10469559 on Build GRCh38
UniGene ID
Hs.711141
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017533.2 NP_060003.2 31 4447
XM_017024676.1 XP_016880165.1 29 4591
GenBank mRNA AF111783.2 - 31 4447

View Full Product Details